missing translation for 'onlineSavingsMsg'
Learn More

Isophthalaldehyde, 98%

Product Code. 11482448
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11482448 5 g 5g
11492448 25 g 25g
11402458 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11482448 Supplier Thermo Scientific Alfa Aesar Supplier No. A15788.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 626-19-7 | C8H6O2 | 134.134 g/mol

Isophthalaldehyde is used in the synthesis of binuclear ruthenium complex. It participates in base-catalyzed Knoevenagel condensation reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isophthalaldehyde is used in the synthesis of binuclear ruthenium complex. It participates in base-catalyzed Knoevenagel condensation reaction.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-19-7
Molecular Formula C8H6O2
Molecular Weight (g/mol) 134.134
MDL Number MFCD00003372
InChI Key IZALUMVGBVKPJD-UHFFFAOYSA-N
Synonym isophthalaldehyde, m-phthalaldehyde, 1,3-benzenedicarboxaldehyde, 1,3-benzenedialdehyde, isophtaldehydes, isophthaldehyde, isophthaldialdehyde, isophtaldehydes french, isophthalic dicarboxaldehyde, unii-lu162b2n9x
PubChem CID 34777
IUPAC Name benzene-1,3-dicarbaldehyde
SMILES C1=CC(=CC(=C1)C=O)C=O

Specifications

Melting Point 85°C to 91°C
Boiling Point 136°C (17 mmHg)
Quantity 5 g
Beilstein 1561038
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 134.13
Percent Purity 98%
Chemical Name or Material Isophthalaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.