missing translation for 'onlineSavingsMsg'
Learn More

Isophorone Diamine-d5 (Major) Dihydrochloride Salt (cis/trans Mixture), TRC

Product Code. 30389370
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389370 25 mg 25mg
30395452 1 mg -
30401108 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30389370 Supplier Toronto Research Chemicals Supplier No. TRCI82193725MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1794752-32-1
Molecular Formula C10 D5 H17 N2 . 2 H Cl
Molecular Weight (g/mol) 248.25
Synonym 5-Amino-1,3,3-trimethylcyclohexanemethanamine-d5 Dihydrochloride, 5-Amino-1,3,3-trimethyl- cyclohexanemethylamine-d5 Dihydrochloride, 5-Amino-1,3,3-trimethylcyclohexanemethylamine-d5 Dihydrochloride, Araldite HY 5161-d5 Dihydrochloride, Chemammina CA 17-d5 Dihydrochloride, D 230-d5, Epikure Dihydrochloride 943-d5, Epilox H 10-31-d5, Epilox IPD-d5, HY 5161-d5, IPD-d5, LH 281-d5, Luxam IPD-d5, Polypox IPD-d5, RIM 137-d5, Rutadur SG-d5 Dihydrochloride, Rutapox H 550L-d5 Dihydrochloride, Vestamin IPD-d5 Dihydrochloride, Vestamine IPDA-d5 Dihydrochloride
IUPAC Name 3-(aminomethyl)-4,4-dideuterio-5,5-dimethyl-3-(trideuteriomethyl)cyclohexanamine;dihydrochloride
SMILES Cl.Cl.[2H]C([2H])([2H])C1(CN)CC(N)CC(C)(C)C1([2H])[2H]

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C10H22N2.2ClH/c1-9(2)4-8(12)5-10(3,6-9)7-11;;/h8H,4-7,11-12H2,1-3H3;2*1H/i3D3,6D2;;
Formula Weight 247.163
Percent Purity >95%
Chemical Name or Material rac-Isophorone Diamine-d5 dihydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.