missing translation for 'onlineSavingsMsg'
Learn More

Isophorone Diamine (cis/trans Mixture), TRC

Product Code. 30405306
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30405306 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30405306 Supplier Toronto Research Chemicals Supplier No. TRCI8219355G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isophorone Diamine
CAS 2855-13-2
Molecular Formula C10 H22 N2
Quantity 5 g
Synonym Cyclohexanemethylamine, 5-amino-1,3,3-trimethyl- (7CI,8CI), 5-Amino-1,3,3-trimethylcyclohexanemethanamine, 1,3,3-Trimethyl-1-aminomethyl-5-aminocyclohexane, 1-Amino-3,3,5-trimethyl-5-aminomethylcyclohexane, 1-Amino-3-(aminomethyl)-3,5,5-trimethylcyclohexane, 3,3,5-Trimethyl-5-aminomethylcyclohexylamine, 3-(Aminomethyl)-3,5,5-trimethylcyclohexanamine, 3-Amino-1,5,5-trimethyl-1-cyclohexanemathanamine, 3-Aminomethyl-3,5,5-trimethyl-1-cyclohexylamine, 3-Aminomethyl-3,5,5-trimethylcyclohexylamine, 5-Amino-1,3,3-trimethylcyclohexanemethylamine, Aradur 42, Araldite HY 5083, Araldite HY 5161, Baxxodur EC 201, CeTePox 1314NFN, Chemammina CA 17, Chemcure 331, D 230, EPH 943, Epicure 3300, Epikure 3300, Epikure 943, Epilox H 10-31, Epilox IPD, HY 3484, HY 5161, IPD, IPDA, Isophorondiamine, Isophorone diamine, LH 281, Luxam IPD, Polypox IPD, RIM 137, Rutadur SG, Rutapox H 550L, Vestamid IPD, Vestamin IPD, Vestamine IPDA, Vestanat IPD
InChI Formula InChI=1S/C10H22N2/c1-9(2)4-8(12)5-10(3,6-9)7-11/h8H,4-7,11-12H2,1-3H3
SMILES CC1(C)CC(N)CC(C)(CN)C1
IUPAC Name 3-(aminomethyl)-3,5,5-trimethylcyclohexan-1-amine
Molecular Weight (g/mol) 170.3
Formula Weight 170.1783
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.