missing translation for 'onlineSavingsMsg'
Learn More

Isophorone, 97%, Thermo Scientific Chemicals

Product Code. 10518860
Click to view available options
Quantity:
5 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-59-1
Molecular Formula C9H14O
Molecular Weight (g/mol) 138.21
MDL Number MFCD00001584
InChI Key HJOVHMDZYOCNQW-UHFFFAOYSA-N
Synonym isophorone, isoacetophorone, isoforone, isoforon, izoforon, isooctopherone, 3,5,5-trimethyl-2-cyclohexen-1-one, alpha-isophorone, 2-cyclohexen-1-one, 3,5,5-trimethyl, 1,1,3-trimethyl-3-cyclohexene-5-one
PubChem CID 6544
ChEBI CHEBI:34800
IUPAC Name 3,5,5-trimethylcyclohex-2-en-1-one
SMILES CC1=CC(=O)CC(C)(C)C1

Specifications

Melting Point -8.0°C
Color Colorless to Yellow
Density 0.9200g/mL
Boiling Point 213.0°C to 214.0°C
Flash Point 84°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4770 to 1.4790
Quantity 5 mL
Beilstein 07,65
Merck Index 15,5242
Specific Gravity 0.92
Solubility Information Solubility in water: 12g/L (20°C). Other solubilities: soluble in alcohol,ether,acetone
Viscosity 2.6 mPa.s (20°C)
Formula Weight 138.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Isophorone, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Isophorone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'