missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopeptidase T/USP5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | Isopeptidase T/USP5 |
|---|---|
| Dilution | Western Blot 1:1000 - 1:2000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30230306
|
Novus Biologicals
NBP3-38080-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30228330
|
Novus Biologicals
NBP3-38080-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Isopeptidase T/USP5 Polyclonal antibody specifically detects Isopeptidase T/USP5 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ImmunofluorescenceSpecifications
| Isopeptidase T/USP5 | |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Deubiquitinating enzyme 5, EC 3.1.2.15, EC 3.4.19.12, Isopeptidase T, IsoT, ISOTubiquitin carboxyl-terminal hydrolase 5, ubiquitin isopeptidase T, ubiquitin specific peptidase 5 (isopeptidase T), ubiquitin specific protease 5 (isopeptidase T), ubiquitin thioesterase 5, Ubiquitin thiolesterase 5, ubiquitin-specific protease-5 (ubiquitin isopeptidase T), Ubiquitin-specific-processing protease 5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human Isopeptidase T/USP5 (NP_003472.2).,, Sequence:, MAELSEEALLSVLPTIRVPKAGDRVHKDECAFSFDTPESEGGLYICMNTFLGFGKQYVERHFNKTGQRVYLHLRRTRRPKEEDPATGTGDPPRKKPTRLAIGVEGGFDLSEEKFELDEDVKIVILPDYLEIARDGLGGLPDIVRDRVTSAVEALLSADSASRKQEVQAWDGEVRQVSKHAFSLKQLDNPARIPPCGWKCSKCDMRENLWLNLTDGSILCGRRYFDGSGGNNHAVEHYRETGYPLAVKLGTITPDGADVYSYDEDDMVLDPSLAEHLSHFG | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:1000 - 1:2000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 8078 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title