missing translation for 'onlineSavingsMsg'
Learn More

Isopentyltriphenylphosphonium bromide, 98%, Thermo Scientific™

Product Code. 11423724
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423724 50 g 50g
11433724 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11423724 Supplier Thermo Scientific Alfa Aesar Supplier No. A11322.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28322-40-9
Molecular Formula C23H26BrP
Molecular Weight (g/mol) 413.34
MDL Number MFCD00031594
InChI Key GZLGTVRDLCJQTO-UHFFFAOYSA-M
Synonym isopentyltriphenylphosphonium bromide, isoamyltriphenylphosphonium bromide, isoamyl triphenylphosphonium bromide, 3-methylbutyl triphenylphosphanium bromide, 3-methylbutyl triphenylphosphonium bromide, 3-methylbutyl triphenyl phosphanium bromide, 2-isopropoxyethyl triphenylphosphonium bromide, acmc-1cjq9, isoamyltriphenylphosphoniumbromide, iso-amyl triphenylphosphonium bromide
PubChem CID 2759379
SMILES [Br-].CC(C)CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∼155°C
Quantity 50 g
Beilstein 3804221
Sensitivity Hygroscopic
IUPAC Name (3-methylbutyl)triphenylphosphanium bromide
Formula Weight 413.35
Percent Purity 98%
Chemical Name or Material Isopentyltriphenylphosphonium bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.