missing translation for 'onlineSavingsMsg'
Learn More

Isopentyl Pentyl Phthalate, TRC

Product Code. 30397688
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30397688 50 mg
30397562 25 mg
30392341 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30397688 Supplier Toronto Research Chemicals Supplier No. TRCI82193050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 776297-69-9
Molecular Weight (g/mol) 306.4
Synonym 1,2-Benzenedicarboxylic acid, 1-(3-methylbutyl) 2-pentyl ester, 1,2-Benzenedicarboxylic acid, 3-methylbutyl pentyl ester (9CI), 1-(3-Methylbutyl) 2-pentyl 1,2-benzenedicarboxylate, n-Pentyl-isopentylphthalate, Isopentyl pentyl phthalate, Phthalic acid, n-pentyl-isopentyl ester
IUPAC Name 2-O-(3-methylbutyl) 1-O-pentyl benzene-1,2-dicarboxylate
SMILES CCCCCOC(=O)c1ccccc1C(=O)OCCC(C)C

Specifications

Molecular Formula C18 H26 O4
Quantity 50 mg
InChI Formula InChI=1S/C18H26O4/c1-4-5-8-12-21-17(19)15-9-6-7-10-16(15)18(20)22-13-11-14(2)3/h6-7,9-10,14H,4-5,8,11-13H2,1-3H3
Formula Weight 306.1831
Percent Purity >95%
Chemical Name or Material Isopentyl Pentyl Phthalate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.