missing translation for 'onlineSavingsMsg'
Learn More

Isopentyl Nitrite, TRC

Product Code. 30391435
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
missing translation for 'unitSize'
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391435 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30391435 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCI821890100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isopentyl Nitrite
CAS 110-46-3
Molecular Formula C5 H11 N O2
Quantity 100 g
Synonym Nitrous acid, 3-methylbutyl ester, Isopentyl alcohol, nitrite (8CI), 3-Methylbutanol nitrite, 3-Methylbutyl nitrite, Amyl nitrite, Aspiral, Isoamyl nitrite, Isopentyl nitrite, NSC 7903, Nitramyl, Nitrous acid, isopentyl ester, Pentyl nitrite, Vaporole
InChI Formula InChI=1S/C5H11NO2/c1-5(2)3-4-8-6-7/h5H,3-4H2,1-2H3
SMILES CC(C)CCON=O
IUPAC Name 3-methylbutyl nitrite
Molecular Weight (g/mol) 117.15
Formula Weight 117.079
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.