missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Isopentyl mercaptan, 97%

Product Code. 11382187
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11382187 100 mL 100mL
11372187 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382187 Supplier Alfa Aesar™ Supplier No. L09661.AE

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 541-31-1
Molecular Formula C5H12S
Molecular Weight (g/mol) 104.211
MDL Number MFCD00004899
InChI Key GIJGXNFNUUFEGH-UHFFFAOYSA-N
Synonym isoamyl mercaptan, 3-methyl-1-butanethiol, 1-butanethiol, 3-methyl, isopentyl mercaptan, 3-methylbutanethiol, isoamylthiol, isopentanethiol, isoamylmercaptan, unii-mmk4sun45e, isoamyl sulfhydrate
PubChem CID 10925
IUPAC Name 3-methylbutane-1-thiol
SMILES CC(C)CCS

Specifications

Density 0.835
Boiling Point 117°C to 118°C
Flash Point 19°C (66°F)
Refractive Index 1.443
Quantity 100 mL
UN Number UN1111
Beilstein 635659
Sensitivity Air Sensitive
Formula Weight 104.22
Percent Purity 97%
Chemical Name or Material Isopentyl mercaptan
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.