missing translation for 'onlineSavingsMsg'
Learn More

Isopentyl isobutyrate, 98%, Thermo Scientific™

Product Code. 11441337
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11441337

Brand: Thermo Scientific Alfa Aesar B24879.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2050-01-3
MDL Number MFCD00053719

Specifications

Density 0.857
Boiling Point 162°C
Flash Point 56°C (133°F)
Molecular Formula C19H18O3
Linear Formula (CH3)2CHCH2CH2O2CCH(CH3)2
Refractive Index 1.407
Quantity 500 g
UN Number UN2620
Synonym 1e,4e-1,5-bis 4-methoxyphenyl penta-1,4-dien-3-one, di-p-anisylideneacetone, bis 4-methoxybenzylidene acetone, 1,5-bis 4-methoxyphenyl penta-1,4-dien-3-one, 1,5-di 4-methoxyphenyl penta-1,4-dien-3-one, trans,trans-1,5-bis 4-methoxyphenyl-1,4-pentadien-3-one, 1e,4e-1,5-bis 4-methoxyphenyl-1,4-pentadien-3-one, dianisylideneacetone, 1,4-pentadien-3-one, 1,5-bis 4-methoxyphenyl, di 4-methoxystyryl ketone
InChI Key IOZVKDXPBWBUKY-LQIBPGRFSA-N
SMILES COC1=CC=C(C=C1)C=CC(=O)C=CC2=CC=C(C=C2)OC
IUPAC Name (1E,4E)-1,5-bis(4-methoxyphenyl)penta-1,4-dien-3-one
Molecular Weight (g/mol) 294.35
PubChem CID 715840
Formula Weight 158.24
Percent Purity 98%
Chemical Name or Material Isopentyl isobutyrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.