missing translation for 'onlineSavingsMsg'
Learn More

Isopentyl butyrate, 98%

Product Code. 11424567
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11424567 500 g 500g
11414567 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11424567 Supplier Thermo Scientific Alfa Aesar Supplier No. B21103.36
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 106-27-4 | C9H18O2 | 158.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-27-4
Molecular Formula C9H18O2
Molecular Weight (g/mol) 158.24
MDL Number MFCD00044888
InChI Key PQLMXFQTAMDXIZ-UHFFFAOYSA-N
Synonym isoamyl butyrate, isopentyl butyrate, isopentyl butanoate, isoamyl butanoate, butanoic acid, 3-methylbutyl ester, 3-methylbutyl butyrate, butyric acid, isopentyl ester, isoamyl-n-butyrate, isopentyl alcohol, butyrate, isoamyl butylate
PubChem CID 7795
ChEBI CHEBI:87422
IUPAC Name 3-methylbutyl butanoate
SMILES CCCC(=O)OCCC(C)C

Specifications

Melting Point -73°C
Density 0.88
Boiling Point 178°C
Flash Point 59°C (138°F)
Odor Fruit-like
Refractive Index 1.411
Quantity 500 g
UN Number UN2620
Merck Index 14,5115
Formula Weight 158.24
Percent Purity 98%
Chemical Name or Material Isopentyl butyrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.