missing translation for 'onlineSavingsMsg'
Learn More

Isopentyl Acetate, Reagent grade, 98%, Honeywell™

Product Code. 15651040 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
20 L
Packaging:
Glass Bottle
Stainless Steel Can
Unit Size:
100mL
20 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15651040 500 mL Glass Bottle 500mL
15631040 100 mL Glass Bottle 100mL
15641040 20 L Stainless Steel Can 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15651040 Supplier Honeywell Research Chemicals Supplier No. 112674500ML

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent Grade, 98%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-92-2
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
MDL Number MFCD00008946
InChI Key MLFHJEHSLIIPHL-UHFFFAOYSA-N
Synonym isoamyl acetate, isopentyl acetate, isopentyl ethanoate, banana oil, pear oil, 3-methylbutyl ethanoate, isoamyl ethanoate, iso-amyl acetate, 3-methyl-1-butyl acetate, i-amyl acetate
PubChem CID 31276
ChEBI CHEBI:31725
IUPAC Name 3-methylbutyl acetate
SMILES CC(C)CCOC(C)=O

Specifications

Melting Point -78°C
Color Colorless
Density 0.876g/mL at 25°C
Boiling Point 142°C
Explosion Limit 0.075
Flash Point 25°C (77°F)
Ignition Point 680°F
Packaging Glass Bottle
Linear Formula CH3COOCH2CH2CH(CH3)2
Refractive Index n20/D 1.4(lit.)
Quantity 500 mL
UN Number UN1104
Beilstein 1744750
Vapor Density 4.5 (vs air)
Formula Weight 130.18g/mol
Vapor Pressure 5mmHg (25°C)
Percent Purity 98%
Grade Reagent
Physical Form Liquid
Chemical Name or Material Isopentyl Acetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.