missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopentenyl Pyrophosphate Ammonium Salt-13D2 (Contains ~1eq H3PO4), TRC
Specifications
Specifications
| Chemical Name or Material | Isopentenyl Pyrophosphate Ammonium Salt-13C2 |
| Molecular Formula | C313C2H12O7P2 xNH3 |
| Quantity | 2.5 mg |
| Synonym | Diphosphoric Acid Mono(3-methyl-3-butenyl) Ester Ammonium Salt-13C2, Ammonium 3-Methyl-3-butenyl Pyrophosphate-13C2 |
| SMILES | C=C([13CH2][13CH2]OP(OP(O)(O)=O)(O)=O)C |
| Molecular Weight (g/mol) | 248.08 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?