missing translation for 'onlineSavingsMsg'
Learn More

Isooctane Blank, VHG™

Product Code. 30003129
Click to view available options
Quantity:
500 mL
0.5 gal
Unit Size:
0.5 GAL
500mL
This item is not returnable. View return policy

Product Code. 30003129

Brand: VHG Labs VHGISO1/2GAL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Isooctane Blank
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,2,4-Trimethylpentane
CAS 540-84-1
Methods ICP/XRF/RDE
Analyte or Component Names 2,2,4-Trimethylpentane
Shipping Condition Room Temperature
Molecular Formula C8 H18
Synonym Pentane, 2,2,4-trimethyl-, 2,2,4-Trimethylpentane, 2,4,4-Trimethylpentane, Isobutyltrimethylmethane, Isooctane, NSC 39117, iso-Octane
InChI Formula InChI=1S/C8H18/c1-7(2)6-8(3,4)5/h7H,6H2,1-5H3
SMILES CC(C)CC(C)(C)C
IUPAC Name 2,2,4-trimethylpentane
Solvent or Matrix Isooctane
Molecular Weight (g/mol) 114.23
Quantity 0.5 gal
Formula Weight 114.1409 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.