missing translation for 'onlineSavingsMsg'
Learn More

Isonitrosoacetophenone, 97%, Thermo Scientific™

Product Code. 11381507
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11381507 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11381507 Supplier Thermo Scientific Alfa Aesar Supplier No. L02098.18

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 532-54-7
Molecular Formula C8H7NO2
Molecular Weight (g/mol) 149.149
MDL Number MFCD00002122
InChI Key MLNKXLRYCLKJSS-RMKNXTFCSA-N
Synonym isonitrosoacetophenone, 2-isonitrosoacetophenone, benzoylformaldoxime, inaf, phenylglyoxal aldoxime, phenylglyoxal 2-oxime, 2-hydroxyiminoacetophenone, phenylglyoxal monoxime, acetophenone, 2-hydroxyimino, phenylglyoxaldoxime
PubChem CID 9566037
IUPAC Name (2E)-2-hydroxyimino-1-phenylethanone
SMILES C1=CC=C(C=C1)C(=O)C=NO

Specifications

Melting Point 124°C to 128°C
Quantity 50 g
Beilstein 2041691
Merck Index 14,5191
Formula Weight 149.15
Percent Purity 97%
Chemical Name or Material Isonitrosoacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.