missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isonipecotic Acid Methyl Ester-13C3 Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Isonipecotic Acid Methyl Ester-13C3 Hydrochloride |
| Molecular Formula | 13C3 C4 H13 N O2 . H Cl |
| Quantity | 5 mg |
| Synonym | 4-Piperidinecarboxylic Acid Methyl Ester-13C3 Hydrochloride, 4-(Methoxycarbonyl)piperidine-13C3 Hydrochloride, Methyl 4-Piperidinecarboxylate-13C3 Hydrochloride, Methyl Isonipecotate-13C3 Hydrochloride, Methyl Isonipecotinate-13C3 Hydrochloride |
| InChI Formula | InChI=1S/C7H13NO2.ClH/c1-10-7(9)6-2-4-8-5-3-6;/h6,8H,2-5H2,1H3;1H/i4+1,5+1,7+1; |
| SMILES | Cl.CO[13C](=O)C1C[13CH2]N[13CH2]C1 |
| IUPAC Name | methyl (2,6-13C2)azinane-4-(13C)carboxylate;hydrochloride |
| Molecular Weight (g/mol) | 182.623 |
| Formula Weight | 182.081 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?