missing translation for 'onlineSavingsMsg'
Learn More

Isonipecotic acid, 98%

Product Code. 11409383
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409383 100 g 100g
11499373 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11409383 Supplier Thermo Scientific Alfa Aesar Supplier No. A11795.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 498-94-2 | C6H11NO2 | 129.16 g/mol

A GABA-A receptor partial agonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 498-94-2
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.16
MDL Number MFCD00006004
InChI Key SRJOCJYGOFTFLH-UHFFFAOYSA-N
Synonym isonipecotic acid, 4-piperidinecarboxylic acid, 4-carboxypiperidine, hexahydroisonicotinic acid, dl-isopipecolinic acid, isonicotinic acid, hexahydro, h-inp-oh, acide isonipecotique, 4-hexahydroisonicotinic acid, unii-m5tzp1rwie
PubChem CID 3773
IUPAC Name piperidine-4-carboxylic acid
SMILES OC(=O)C1CCNCC1

Specifications

Melting Point >350°C
Odor Odorless
Quantity 100 g
Beilstein 112553
Merck Index 14,5189
Formula Weight 129.16
Percent Purity 98%
Chemical Name or Material Isonipecotic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.