missing translation for 'onlineSavingsMsg'
Learn More

Isonipecotamide, 98%

Product Code. 11485493
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11485493 10 g 10g
11495493 50 g 50g
11405503 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11485493 Supplier Thermo Scientific Alfa Aesar Supplier No. A13442.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 39546-32-2 | C6H12N2O | 128.18 g/mol

Isonipecotamide is used as a reactant for the preparation of Survival motor neuron (SMN) protein modulators, orally available naphthyridine protein kinase D inhibitors and heteroalicyclic carboxamidines as inhibitors of inducible nitric oxide synthase. It is also used to prepare phosphodiesterase 5 inhibitors. It is also used as a reactant for identification of small molecular inhibitors of importin-beta mediated nuclear import. It is also used as a fluorenylmethyloxycarbonyl - deprotection agent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isonipecotamide is used as a reactant for the preparation of Survival motor neuron (SMN) protein modulators, orally available naphthyridine protein kinase D inhibitors and heteroalicyclic carboxamidines as inhibitors of inducible nitric oxide synthase. It is also used to prepare phosphodiesterase 5 inhibitors. It is also used as a reactant for identification of small molecular inhibitors of importin-beta mediated nuclear import. It is also used as a fluorenylmethyloxycarbonyl - deprotection agent in organic synthesis.

Solubility
Soluble in water.

Notes
Moisture sensitive. Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39546-32-2
Molecular Formula C6H12N2O
Molecular Weight (g/mol) 128.18
MDL Number MFCD00038012
InChI Key DPBWFNDFMCCGGJ-UHFFFAOYSA-N
Synonym isonipecotamide, 4-piperidinecarboxamide, hexahydroisonicotinamide, 4-carbamoylpiperidine, isonipectoamide, piperidine-4-carboxylic acid amide, 4-piperdinecarboxamide, piperidin-4-carboxamid, iso nipecotamide, iso-nipecotamide
PubChem CID 3772
IUPAC Name piperidine-4-carboxamide
SMILES NC(=O)C1CCNCC1

Specifications

Melting Point 148°C to 149°C
Quantity 10 g
Beilstein 112554
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 128.18
Percent Purity 98%
Chemical Name or Material Isonipecotamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.