missing translation for 'onlineSavingsMsg'
Learn More

Isonicotinoyl chloride hydrochloride, 97%

Product Code. 10043813 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Packaging:
Plastic bottle
Unit Size:
100g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10043813 250 g Plastic bottle 250g
10012680 25 g Plastic bottle 25g
10762251 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10043813 Supplier Thermo Scientific Acros Supplier No. 211682500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 39178-35-3 | C6H5Cl2NO | 178.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39178-35-3
Molecular Formula C6H5Cl2NO
Molecular Weight (g/mol) 178.01
MDL Number MFCD00012830
InChI Key BNTRVUUJBGBGLZ-UHFFFAOYSA-N
Synonym isonicotinoyl chloride hydrochloride, pyridine-4-carbonyl chloride hydrochloride, isonicotinoylchloride hydrochloride, 4-pyridinecarbonyl chloride hydrochloride, 4-pyridinecarbonyl chloride, hydrochloride, isonicotinoylchloridehcl, pubchem23974, acmc-1bn6p, ksc223m5d, 4-pyridinecarbonyl chloride-hcl
PubChem CID 12262826
IUPAC Name pyridine-4-carbonyl chloride;hydrochloride
SMILES [H+].[Cl-].ClC(=O)C1=CC=NC=C1

Specifications

Melting Point 153.0°C to 161.0°C
Color Beige to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Plastic bottle
Quantity 250 g
Beilstein 22, 46
Formula Weight 178.02
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Isonicotinoyl chloride hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.