missing translation for 'onlineSavingsMsg'
Learn More

Isonicotinic Acid, TRC

Product Code. 30389947
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30389947 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30389947 Supplier Toronto Research Chemicals Supplier No. TRCI821760100G
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pyridine-4-carboxylic Acid (Isonicotinic Acid)
CAS 55-22-1
Purity Grade Notes HPLC
Molecular Formula C6 H5 N O2
Quantity 100 g
Synonym Pyridine-4-carboxylic Acid, Nicotinic Acid Imp. E (EP), Isonicotinic Acid, Pyridine-4-carboxylic Acid, Nicotinic Acid Imp. E (EP), Isonicotinic Acid, Isoniazid Impurity A, Nicotinic Acid Impurity E
InChI Formula InChI=1S/C6H5NO2/c8-6(9)5-1-3-7-4-2-5/h1-4H,(H,8,9)
SMILES OC(=O)c1ccncc1
IUPAC Name pyridine-4-carboxylic acid
Molecular Weight (g/mol) 123.11
Formula Weight 123.032
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.