missing translation for 'onlineSavingsMsg'
Learn More

Isonicotinamide, 99%

Product Code. 11423717
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
500 g
Unit Size:
100g
250g
500g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423717 50 g 50g
11438870 100 g 100g
11433717 250 g 250g
11448870 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11423717 Supplier Thermo Scientific Alfa Aesar Supplier No. A18190.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1453-82-3 | C6H6N2O | 122.127 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1453-82-3
Molecular Formula C6H6N2O
Molecular Weight (g/mol) 122.127
MDL Number MFCD00006432
InChI Key VFQXVTODMYMSMJ-UHFFFAOYSA-N
Synonym isonicotinamide, 4-pyridinecarboxamide, 4-carbamoylpyridine, isonicotinic acid amide, isonicotineamide, gamma-pyridinecarboxamide, pyridine-4-carboxylic acid amide, unii-4h3bh6yx9q, .gamma.-pyridinecarboxamide, 4h3bh6yx9q
PubChem CID 15074
ChEBI CHEBI:6031
IUPAC Name pyridine-4-carboxamide
SMILES C1=CN=CC=C1C(=O)N

Specifications

Melting Point 155°C to 158°C
Quantity 50 g
Beilstein 2173
Formula Weight 122.13
Percent Purity 99%
Chemical Name or Material Isonicotinamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.