missing translation for 'onlineSavingsMsg'
Learn More

Isoniazid, MedChemExpress

Product Code. 16478236
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478236 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16478236 Supplier Medchem Express Supplier No. HYB0329
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Isoniazid (INH) is a prodrug and must be activated by a bacterial catalase-peroxidase enzyme KatG. Isoniazid is bactericidal to rapidly dividing mycobacteria and has anti-tuberculostatic activity.

Isoniazid (INH) is a prodrug and must be activated by a bacterial catalase-peroxidase enzyme KatG. Isoniazid is bactericidal to rapidly dividing mycobacteria and has anti-tuberculostatic activity.

Specifications

Chemical Name or Material Isoniazid
Physical Form Solid
CAS 54-85-3
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C6H7N3O
For Use With (Application) COVID-19-immunoregulation
Synonym INH Isonicotinic acid hydrazide Isonicotinic hydrazide
Solubility Information DMSO : 50 mg/mL (364.59 mM; Need ultrasonic) ∣H2O : 33.33 mg/mL (243.04 mM; Need ultrasonic)
SMILES O=C(C1=CC=NC=C1)NN
Molecular Weight (g/mol) 137.14
Formula Weight 137.14
Percent Purity 99.68%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.