missing translation for 'onlineSavingsMsg'
Learn More

Isomaltotriose, TRC

Product Code. 30397160
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397160 25 mg 25mg
30400803 50 mg -
30404153 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30397160 Supplier Toronto Research Chemicals Supplier No. TRCI82129825MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3371-50-4
Molecular Formula C18H32O16
Molecular Weight (g/mol) 504.44
Synonym O-α-D-glucopyranosyl-(1→6)-O-α-D-glucopyranosyl-(1→6)-D-Glucose, 6-α-Isomaltosylglucose
IUPAC Name (3R,4S,5S,6R)-6-((((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-((((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-2,3,4,5-tetraol
SMILES C([C@@H]1[C@H]([C@@H]([C@H]([C@H](O1)OC[C@@H]2[C@H]([C@@H]([C@H]([C@H](O2)OC[C@@H]3[C@H]([C@@H]([C@H](C(O3)O)O)O)O)O)O)O)O)O)O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C18H32O16/c19-1-4-7(20)11(24)14(27)17(33-4)31-3-6-9(22)12(25)15(28)18(34-6)30-2-5-8(21)10(23)13(26)16(29)32-5/h4-29H,1-3H2/t4-,5-,6-,7-,8-,9-,10+,11+,12+,13-,14-,15-,16?,17+,18+/m1/s1
Formula Weight 504.17
Percent Purity >95%
Chemical Name or Material Isomaltotriose
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.