missing translation for 'onlineSavingsMsg'
Learn More

Isoleucylleucine TFA Salt, TRC

Product Code. 30397139
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397139 500 mg -
30389345 100 mg -
30398804 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30397139 Supplier Toronto Research Chemicals Supplier No. TRCI416275500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 160448-53-3
Molecular Formula C12H24N2O3 . C2HF3O2
Molecular Weight (g/mol) 358.35
Synonym L-Isoleucyl-L-leucine TFA Salt, 193 TFA Salt: PN TFA Salt: EP2071334 SEQID TFA Salt: 210 Claimed Protein TFA Salt, 193 TFA Salt: PN: WO2009077864 SEQID: 210 Claimed Protein TFA Salt
IUPAC Name (2S)-2-[[(2S,3S)-2-amino-3-methylpentanoyl]amino]-4-methylpentanoic acid;2,2,2-trifluoroacetic acid
SMILES CC[C@H](C)[C@H](N)C(=O)N[C@@H](CC(C)C)C(=O)O.OC(=O)C(F)(F)F

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C12H24N2O3.C2HF3O2/c1-5-8(4)10(13)11(15)14-9(12(16)17)6-7(2)3;3-2(4,5)1(6)7/h7-10H,5-6,13H2,1-4H3,(H,14,15)(H,16,17);(H,6,7)/t8-,9-,10-;/m0./s1
Formula Weight 358.1716
Chemical Name or Material Isoleucylleucine TFA Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.