missing translation for 'onlineSavingsMsg'
Learn More

Isoleucine O-Benzyl Valsartan, TRC

Product Code. 30391902
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391902 5 mg 5mg
30389886 25 mg 25mg
30392623 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30391902 Supplier Toronto Research Chemicals Supplier No. TRCI8222305MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isoleucine O-Benzyl Valsartan
Analyte or Component Names Isoleucine O-Benzyl Valsartan
Shipping Condition Room Temperature
Molecular Formula C32 H37 N5 O3
Synonym (2S,3S)-Benzyl 2-(N-((2'-(1H-Tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)pentanamido)-3-methylpentanoate
InChI Formula InChI=1S/C32H37N5O3/c1-4-6-16-29(38)37(30(23(3)5-2)32(39)40-22-25-12-8-7-9-13-25)21-24-17-19-26(20-18-24)27-14-10-11-15-28(27)31-33-35-36-34-31/h7-15,17-20,23,30H,4-6,16,21-22H2,1-3H3,(H,33,34,35,36)/t23-,30-/m0/s1
Solution Type Neat
SMILES CCCCC(=O)N(Cc1ccc(cc1)c2ccccc2c3nnn[nH]3)[C@@H]([C@@H](C)CC)C(=O)OCc4ccccc4
IUPAC Name benzyl (2S,3S)-3-methyl-2-[pentanoyl-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]amino]pentanoate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 539.668
Quantity 5 mg
Formula Weight 539.29
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.