missing translation for 'onlineSavingsMsg'
Learn More

Isoleucine O-Benzyl N-Titryl Valsartan, TRC

Product Code. 30390790
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390790 25 mg 25mg
30405887 10 mg 10mg
30389371 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390790 Supplier Toronto Research Chemicals Supplier No. TRCI82220525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isoleucine O-Benzyl N-Titryl Valsartan
Analyte or Component Names Isoleucine O-Benzyl N-Titryl Valsartan
Shipping Condition Room Temperature
Molecular Formula C51 H51 N5 O3
Synonym (2S,3S)-Benzyl 3-Methyl-2-(N-((2'-(2-trityl-2H-tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)pentanamido)pentanoate
InChI Formula InChI=1S/C51H51N5O3/c1-4-6-31-47(57)55(48(38(3)5-2)50(58)59-37-40-21-11-7-12-22-40)36-39-32-34-41(35-33-39)45-29-19-20-30-46(45)49-52-54-56(53-49)51(42-23-13-8-14-24-42,43-25-15-9-16-26-43)44-27-17-10-18-28-44/h7-30,32-35,38,48H,4-6,31,36-37H2,1-3H3/t38-,48-/m0/s1
Solution Type Neat
SMILES CCCCC(=O)N(Cc1ccc(cc1)c2ccccc2c3nnn(n3)C(c4ccccc4)(c5ccccc5)c6ccccc6)[C@@H]([C@@H](C)CC)C(=O)OCc7ccccc7
IUPAC Name benzyl (2S,3S)-3-methyl-2-[pentanoyl-[[4-[2-(2-trityltetrazol-5-yl)phenyl]phenyl]methyl]amino]pentanoate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 781.982
Quantity 25 mg
Formula Weight 781.399
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.