missing translation for 'onlineSavingsMsg'
Learn More

Isoimperatorin, MedChemExpress

Product Code. 16403856
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16403856 25 mg 25mg
16448938 10 mg 10mg
16421378 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16403856 Supplier Medchem Express Supplier No. HYN0286

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Isoimperatorin is a methanolic extract of the roots of Angelica dahurica shows significant inhibitory effects on acetylcholinesterase (AChE) with the IC50 of 74.6 μM.

Isoimperatorin is a methanolic extract of the roots of Angelica dahurica shows significant inhibitory effects on acetylcholinesterase (AChE) with the IC50 of 74.6 μM.

Specifications

Chemical Name or Material Isoimperatorin
CAS 482-45-1
Color White
Purity Grade Notes Research
Molecular Formula C16H14O4
Quantity 25 mg
Solubility Information Ethanol : 8.33 mg/mL (30.82 mM; Need ultrasonic) ∣DMSO : 2.5 mg/mL (9.25 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C1C=CC2=C(OC/C=C(C)/C)C3=C(OC=C3)C=C2O1
Molecular Weight (g/mol) 270.28
Formula Weight 270.28
Percent Purity 98.93%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.