Product Code. 30117486
Shop All Thermo Scientific Products
Click to view available options
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Unit Size
30117486
1 L
1 L
10025752
2.5 L
2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code.
30117486
Supplier
Thermo Scientific
Supplier No.
383810010
181.90 GBP valid until 2026-12-18 BEST PRICE on Promo! Use promo code "27967" to get your promotional price.
missing translation for 'validMainframeMsg'
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
CAS: 73513-42-5, C6H14, 86.18 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
CAS
73513-42-5
Molecular Formula
C6H14
Molecular Weight (g/mol)
86.18
MDL Number
MFCD00009406
InChI Key
AFABGHUZZDYHJO-UHFFFAOYSA-N
Synonym
isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane
PubChem CID
7892
IUPAC Name
2-methylpentane
SMILES
CCCC(C)C
Specifications
Melting Point
-154.0°C
Density
0.6600g/mL
Boiling Point
60.0°C
Flash Point
-23°C
Linear Formula
CH3 CH2 CH2 CH(CH3 )2
Refractive Index
1.3700 to 1.3720
Quantity
1 L
Specific Gravity
0.66
Solubility Information
Solubility in water: insoluble. Other solubilities: soluble in ethanol
Formula Weight
86.18
Grade
HPLC
Green Features
ACT Ecolabel (Gold)
Chemical Name or Material
Isohexane
Show More
Show Less
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?