Learn More
IsoGel™ Agarose
Product Code. 11577985
Click to view available options
Quantity:
25 g
Unit Size:
Each
Product Code. 11577985
Brand: Lonza 50202
  • For isoelectric focusing (IEF)
  • No measurable EEO
  • Sufficiently rigid to be cast in vertical tubes
  • Gels cast on GelBond™ Film dry down quickly and easily to transparent permanent records
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Cambrex Iso-Gel™ Agarose
Quantity 25 g