missing translation for 'onlineSavingsMsg'
Learn More

Isoflurane, TRC

Product Code. 30007424
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007424 50 g 50g
30008448 1 g 1g
30003373 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.
Product Code. 30007424 Supplier Toronto Research Chemicals Supplier No. TRCI81680050G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26675-46-7
Molecular Formula C3 H2 Cl F5 O
Molecular Weight (g/mol) 184.49
Synonym Ether, 1-chloro-2,2,2-trifluoroethyl difluoromethyl (8CI), 2-Chloro-2-(difluoromethoxy)-1,1,1-trifluoroethane, (±)-Isoflurane, 1-Chloro-2,2,2-trifluoroethyl difluoromethyl ether, Aerrane, Attane, Compound 469, Difluoromethyl 1-chloro-2,2,2-trifluoroethyl ether, Escain, Forane, Forene, HCFE 235da2, IsoFlo, IsoFlo (anesthetic), Isoba, Isoflurane, Isofor, Isoforine, Isorrane, R-E 235da1
IUPAC Name 2-chloro-2-(difluoromethoxy)-1,1,1-trifluoroethane
SMILES FC(F)OC(Cl)C(F)(F)F

Specifications

Quantity 50 g
InChI Formula InChI=1S/C3H2ClF5O/c4-1(3(7,8)9)10-2(5)6/h1-2H
Formula Weight 183.9714
Chemical Name or Material Isoflurane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.