missing translation for 'onlineSavingsMsg'
Learn More

Isoflupredone Acetate, TRC

Product Code. 17963895
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17963895 5 mg 5mg
17932745 100 mg 100mg
17930745 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17963895 Supplier Toronto Research Chemicals Supplier No. TRCI8166105MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 338-98-7
Molecular Formula C23 H29 F O6
Molecular Weight (g/mol) 420.47
Synonym Pregna-1,4-diene-3,20-dione, 21-(acetyloxy)-9-fluoro-11,17-dihydroxy-, (11β)-, Pregna-1,4-diene-3,20-dione, 9-fluoro-11β,17,21-trihydroxy-, 21-acetate (6CI,7CI,8CI), 21-Acetoxy-9-fluoro-11β,17-dihydroxypregna-1,4-diene-3,20-dione, 9-Fluoro-11β,17,21-trihydroxypregna-1,4-diene-3,20-dione 21-acetate, 9-Fluoroprednisolone 21-acetate, 9-Fluoroprednisolone acetate, 9α-Fluoro-11β,17α,21-trihydroxypregna-1,4-diene-3,20-dione 21-acetate, 9α-Fluoroprednisolone 21-acetate, 9α-Fluoroprednisolone acetate, Isoflupredone acetate, NSC 12600, NSC 37977, Predef, Predef R 2X, U 6013
IUPAC Name [2-[(8S,9R,10S,11S,13S,14S,17R)-9-fluoro-11,17-dihydroxy-10,13-dimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
SMILES CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C23H29FO6/c1-13(25)30-12-19(28)22(29)9-7-16-17-5-4-14-10-15(26)6-8-20(14,2)23(17,24)18(27)11-21(16,22)3/h6,8,10,16-18,27,29H,4-5,7,9,11-12H2,1-3H3/t16-,17-,18-,20-,21-,22-,23-/m0/s1
Formula Weight 420.1948
Percent Purity >95%
Chemical Name or Material Isoflupredone Acetate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.