missing translation for 'onlineSavingsMsg'
Learn More

Isoferulic Acid-d3 3-O-Sulfate Disodium Salt, TRC

Product Code. 30406997
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30406997 50 mg 50mg
30397740 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30406997 Supplier Toronto Research Chemicals Supplier No. TRCI81616250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1795140-83-8
Molecular Formula C10H5D3Na2O7S
Molecular Weight (g/mol) 321.23
Synonym 3-[4-(Methoxy-d3)-3-(sulfooxy)phenyl]-2-propenoic Acid Disodium Salt
SMILES [Na+].[Na+].[2H]C([2H])([2H])Oc1ccc(C=CC(=O)[O-])cc1OS(=O)(=O)[O-]

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C10H10O7S.2Na/c1-16-8-4-2-7(3-5-10(11)12)6-9(8)17-18(13,14)15;;/h2-6H,1H3,(H,11,12)(H,13,14,15);;/q;2*+1/p-2/i1D3;;
Chemical Name or Material Isoferulic Acid-d3 3-O-Sulfate Disodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.