missing translation for 'onlineSavingsMsg'
Learn More

Isofenphos-oxon, Honeywell Fluka™

Product Code. 16606213 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16606213 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16606213 Supplier Honeywell Fluka Supplier No. R169610MG
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.

  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isofenphos-oxon
CAS 31120-85-1
CAS Max % ≤ 100%
Melting Point 32°C to 38°C
Synonym Isopropyl 2-((ethoxy(isopropylamino)phosphoryl)oxy)benzoate
Molecular Weight (g/mol) 329.33 g/mol
Quantity 10 mg
Grade Acaricides and Insecticides
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.