missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isofenphos-methyl, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifications
Specifications
| Chemical Name or Material | Isofenphos-methyl |
| CAS | 99675-03-3 |
| CAS Max % | ≤ 100% |
| Density | 1.5221 g/cm3 |
| Flash Point | 100°C |
| Color | Yellow |
| Synonym | 1-Methylethyl 2-[[methoxy[(1-methylethyl)amino]phosphinothioyl]oxy]benzoate |
| Molecular Weight (g/mol) | 331.37 g/mol |
| Quantity | 50 mg |
| Grade | Acaricides and Insecticides |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction