missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isofenphos-d7, TRC
Specifications
Specifications
| Chemical Name or Material | Isofenphos-d7 |
| Molecular Formula | C15 D7 H17 N O4 P S |
| Quantity | 10 mg |
| Synonym | isopropyl Ester O-Ester With O-Ethyl Isopropyl-d7-phosphoramidothioate Salicylic Acid, Isopropyl-d7-Phosphoramidothioic Acid O-Ethyl Ester O-Ester With Isopropyl Salicylate, Amaze-d7, BAY 92114-d7, Bayer 92114-d7, Isophenphos-d7, Oftanol-d7, Pryfon 6-d7, Pyfron 6-d7, SRA 12869-d7, 2-[[Ethoxy[(1-methylethyl-d7)amino]phosphinothioyl]oxy]benzoic Acid 1-Methylethyl Ester |
| InChI Formula | InChI=1S/C15H24NO4PS/c1-6-18-21(22,16-11(2)3)20-14-10-8-7-9-13(14)15(17)19-12(4)5/h7-12H,6H2,1-5H3,(H,16,22)/i2D3,3D3,11D |
| SMILES | [2H]C([2H])([2H])C([2H])(NP(=S)(OCC)Oc1ccccc1C(=O)OC(C)C)C([2H])([2H])[2H] |
| IUPAC Name | propan-2-yl 2-[ethoxy-(1,1,1,2,3,3,3-heptadeuteriopropan-2-ylamino)phosphinothioyl]oxybenzoate |
| Molecular Weight (g/mol) | 352.437 |
| Formula Weight | 352.16 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?