missing translation for 'onlineSavingsMsg'
Learn More

Isodimethoate, TRC

Product Code. 30397931
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397931 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30397931 Supplier Toronto Research Chemicals Supplier No. TRCI81508510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isodimethoate
CAS 3344-11-4
Purity Grade Notes GC
Molecular Formula C5 H12 N O3 P S2
Quantity 10 mg
Synonym O,S-Dimethyl S-(2-(Methylamino)-2-oxoethyl) Phosphorodithioate, 2-Mercapto-N-methyl-acetamide S-Ester with O,S-Dimethyl Phosphorodithioate, O,S-Dimethyl S-[2-(Methylamino)-2-oxoethyl] Ester Phosphorodithioic Acid, O,S-Dimethyl Ester, S-Ester with 2-Mercapto-N-methylacetamide Phosphorodithioic Acid
InChI Formula InChI=1S/C5H12NO3PS2/c1-6-5(7)4-12-10(8,9-2)11-3/h4H2,1-3H3,(H,6,7)
SMILES CNC(=O)CSP(=O)(OC)SC
IUPAC Name 2-[methoxy(methylsulfanyl)phosphoryl]sulfanyl-N-methylacetamide
Molecular Weight (g/mol) 229.26
Formula Weight 228.9996
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.