missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Isocytosine, 99%

Product Code. 15450568
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15450568

Brand: Thermo Scientific Chemicals H54228.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-53-2 | C4H5N3O | 111.10 g/mol

Isocytosine (2-aminouracil) is an isomer of cytosine used in physical chemical studies involving metal complex binding, hydrogen-bonding, and tautomerism and proton transfer effects in nucleobases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isocytosine (2-aminouracil) is an isomer of cytosine used in physical chemical studies involving metal complex binding, hydrogen-bonding, and tautomerism and proton transfer effects in nucleobases.

Solubility
Soluble in DMF, DMSO, hot water, and AcOH.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-53-2
Molecular Formula C4H5N3O
Molecular Weight (g/mol) 111.10
MDL Number MFCD00057557 MFCD00023256
InChI Key XQCZBXHVTFVIFE-UHFFFAOYSA-N
Synonym isocytosine, 2-amino-4-hydroxypyrimidine, 2-aminopyrimidin-4-ol, 2-aminopyrimidin-4 1h-one, 2-amino-4-pyrimdinol, 2-amino-4-pyrimidone, 2-amino-4-oxypyrimidine, 2-aminouracil, 4-pyrimidinol, 2-amino, 4 1h-pyrimidinone, 2-amino
PubChem CID 66950
ChEBI CHEBI:55502
IUPAC Name 2-amino-3,4-dihydropyrimidin-4-one
SMILES NC1=NC=CC(=O)N1

Specifications

Quantity 5 g
Solubility Information Soluble in DMF,DMSO,hot water,and AcOH.
Formula Weight 111.1
Percent Purity 99%
Chemical Name or Material Isocytosine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.