missing translation for 'onlineSavingsMsg'
Learn More

(Isocyanoimino)triphenylphosphorane, 90%

Product Code. 15545886 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15545886 1 g 1g
15555886 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15545886 Supplier Thermo Scientific Acros Supplier No. 452920010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 73789-56-7 | C19H15N2P | 302.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73789-56-7
Molecular Formula C19H15N2P
Molecular Weight (g/mol) 302.32
MDL Number MFCD09038528
InChI Key NIDTXBFHPXMXTR-UHFFFAOYSA-N
Synonym isocyanoimino triphenylphosphorane, isocyanoiminotriphenylphosphorane, isocyano triphenyl-??-phosphanylidene amine, cnnpph3;, ncnpph3;, ph3pnnc;, acmc-209os9, cn-n=pph3;, n-isocyaniminotriphenylphosphorane, n-isocyanoiminotriphenylphosphorane
PubChem CID 11266621
IUPAC Name isocyanoimino(triphenyl)-$l^{5}-phosphane
SMILES [C-]#[N+]N=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 160°C to 161°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (HPLC)
Quantity 1 g
Formula Weight 302.31
Percent Purity 90%
Chemical Name or Material (Isocyanoimino)triphenylphosphorane
compliance-icons
Product Identifier
  • (Isocyanoimino)triphenylphosphorane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.