missing translation for 'onlineSavingsMsg'
Learn More

(-)-Isocorypalmine, MedChemExpress

Product Code. 16411618
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16411618 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16411618 Supplier Medchem Express Supplier No. HYN0927
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress (-)-Isocorypalmine (Tetrahydrocolumbamine), isolated from the crude base fraction of Corydalis chaerophylla, is a dopamine receptor ligand. Recombinant CYP719A21 displays strict substrate specificity and high affinity (Km=4.63 ± 0.71 μM) for (-)-Isocorypalmine.

(-)-Isocorypalmine (Tetrahydrocolumbamine), isolated from the crude base fraction of Corydalis chaerophylla, is a dopamine receptor ligand. Recombinant CYP719A21 displays strict substrate specificity and high affinity (Km=4.63 ± 0.71 μM) for (-)-Isocorypalmine.

Specifications

Chemical Name or Material (-)-Isocorypalmine
CAS 483-34-1
Color White
Purity Grade Notes Research
Molecular Formula C20H23NO4
Quantity 5 mg
Synonym Tetrahydrocolumbamine (S)-Tetrahydrocolumbamine
SMILES OC(C=C1[C@@]2([H])N3CC4=C(C=CC(OC)=C4OC)C2)=C(C=C1CC3)OC
Molecular Weight (g/mol) 341.4
Formula Weight 341.4
Percent Purity 98.64%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.