missing translation for 'onlineSavingsMsg'
Learn More

Isocorynoxeine, MedChemExpress

Product Code. 16411508
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16411508

Brand: Medchem Express HYN0775

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Isocorynoxeine, an isorhynchophylline-related alkaloid, exhibits a dose-dependent inhibition of 5-HT2A receptor-mediated current response with an IC50 of 72.4 μM.

Isocorynoxeine, an isorhynchophylline-related alkaloid, exhibits a dose-dependent inhibition of 5-HT2A receptor-mediated current response with an IC50 of 72.4 μM.

Specifications

Chemical Name or Material Isocorynoxeine
CAS 51014-29-0
Color White
Purity Grade Notes Research
Molecular Formula C22H26N2O4
Quantity 5 mg
Synonym 7-Isocorynoxeine
Solubility Information DMSO : 25 mg/mL (65.37 mM; Need ultrasonic)
SMILES O=C(NC1=C2C=CC=C1)[C@@]32[C@@](C[C@H](/C(C(OC)=O)=C\OC)[C@@H](C=C)C4)([H])N4CC3
Molecular Weight (g/mol) 382.45
Formula Weight 382.45
Percent Purity 99.95%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.