missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isoconazole nitrate, 99%, Thermo Scientific™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Specifications
Specifications
| Melting Point | 181.0°C to 183.0°C |
| Infrared Spectrum | Authentic |
| Quantity | 25 g |
| Formula Weight | 479.14 |
| Percent Purity | 99% |
| Chemical Name or Material | Isoconazole nitrate |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction