missing translation for 'onlineSavingsMsg'
Learn More

Isocitrate Dehydrogenase 1/IDH1 Antibody (IDH1.R132H/7328R), Novus Biologicals™

Rabbit Monoclonal Antibody

£232.00 - £412.00

Specifications

Antigen Isocitrate Dehydrogenase 1/IDH1
Clone IDH1.R132H/7328R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 μg/mL
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18658944
View Documents
Novus Biologicals
NBP3-20633-20ug
20 μg
£232.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18622905
View Documents
Novus Biologicals
NBP3-20633-100ug
100 μg
£412.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Isocitrate Dehydrogenase 1/IDH1 Monoclonal antibody specifically detects Isocitrate Dehydrogenase 1/IDH1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Isocitrate Dehydrogenase 1/IDH1
0.2 mg/ml
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Stem Cell Markers
1mM PBS with 0.05% BSA
3417
IgG κ
Protein A or G purified
IDH1.R132H/7328R
Immunohistochemistry-Paraffin 1-2 μg/mL
Monoclonal
Purified
RUO
Human
Cytosolic NADP-isocitrate dehydrogenase, EC 1.1.1.42, IDCD, IDH, IDP, IDPC, isocitrate dehydrogenase [NADP] cytoplasmic, isocitrate dehydrogenase 1 (NADP+), soluble, NADP(+)-specific ICDH, NADP-dependent isocitrate dehydrogenase, cytosolic, NADP-dependent isocitrate dehydrogenase, peroxisomal, Oxalosuccinate decarboxylase, PICD
Recombinant fragment (around aa 1-200) of human Isocitrate Dehydrogenase 1/IDH1 (exact sequence is proprietary)
Primary
Store at 4&drg;C. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.