missing translation for 'onlineSavingsMsg'
Learn More

Isochroman, 99%

Product Code. p-150829 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10794543

Brand: Thermo Scientific Acros 122540050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 493-05-0 | C9H10O | 134.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 493-05-0
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00006913
InChI Key HEBMCVBCEDMUOF-UHFFFAOYSA-N
Synonym isochroman, isochromane, 3,4-dihydro-1h-2-benzopyran, 1h-2-benzopyran, 3,4-dihydro, 3,4-dihydro-1h-benzo c pyran, pubchem20553, acmc-209kfu, isochroman 10g, ksc237m1l
PubChem CID 96266
ChEBI CHEBI:33225
IUPAC Name 3,4-dihydro-1H-isochromene
SMILES C1CC2=CC=CC=C2CO1

Specifications

Color Colorless to Yellow
Density 1.0600g/mL
Boiling Point 75°C (4.0 Mbar)
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5430 to 1.546
Quantity 5 g
Beilstein 17, 53
Specific Gravity 1.06
Formula Weight 134.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Isochroman, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.