missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ Isochroman, 99%
Description
Applications
Isochroman is used as an organic chemical synthesis intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Isochroman is used as an organic chemical synthesis intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Specifications
| Density | 1.067 |
| Boiling Point | 89°C to 90°C (12 mmHg) |
| Flash Point | 66°C (151°F) |
| Refractive Index | 1.545 |
| Quantity | 50 g |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 134.18 |
| Percent Purity | 99% |
| Chemical Name or Material | Isochroman |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?