missing translation for 'onlineSavingsMsg'
Learn More

Isochenodeoxycholic Acid, TRC

Product Code. 30137882
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137882 50 mg 50mg
17963025 10 mg 10mg
17902305 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137882 Supplier Toronto Research Chemicals Supplier No. TRCI78960050MG
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 566-24-5
Molecular Formula C24H40O4
Molecular Weight (g/mol) 392.57
Synonym (3β,5β,7α)-3,7-Dihydroxycholan-24-oic Acid, 3β,7α-Dihydroxy-5β-cholan-24-oic Acid, 3β,7α-Dihydroxy-5β-cholanic Acid, 3β,7α-Dihydroxy-5β-cholanoic Acid, 3β-Chenodeoxycholic Acid, BAR601, Isochenodeoxycholic Acid;
SMILES O[C@H]1CC[C@@]2(C)[C@@](C[C@@H](O)[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@]4([H])[C@H](C)CCC(O)=O)([H])C1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C24H40O4/c1-14(4-7-21(27)28)17-5-6-18-22-19(9-11-24(17,18)3)23(2)10-8-16(25)12-15(23)13-20(22)26/h14-20,22,25-26H,4-13H2,1-3H3,(H,27,28)/t14-,15+,16+,17-,18+,19+,20-,22+,23+,24-/m1/s1
Formula Weight 392.2927
Percent Purity >95%
Chemical Name or Material Isochenodeoxycholic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.