missing translation for 'onlineSavingsMsg'
Learn More

Isocaryophyllene (80%), TRC

Product Code. 30394153
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394153 1 g 1g
30401160 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30394153 Supplier Toronto Research Chemicals Supplier No. TRCI7894001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-65-0
Molecular Formula C15 H24
Molecular Weight (g/mol) 204.35
Synonym Bicyclo[7.2.0]undec-4-ene, 4,11,11-trimethyl-8-methylene-, (1R,4Z,9S)-, Bicyclo[7.2.0]undec-4-ene, 4,11,11-trimethyl-8-methylene-, (Z)-(1R,9S)-(-)- (8CI), Bicyclo[7.2.0]undec-4-ene, 4,11,11-trimethyl-8-methylene-, [1R-(1R*,4Z,9S*)]-, Isocaryophyllene (6CI,7CI), (1R,4Z,9S)-4,11,11-Trimethyl-8-methylenebicyclo[7.2.0]undec-4-ene, (-)-Isocaryophyllene, (1R,9S,Z)-4,11,11-Trimethyl-8-methylenebicyclo[7.2.0]undec-4-ene, (Z)-Caryophyllene, Isocaryophyllene, (-)-, cis-Caryophyllene, γ-Caryophyllen
IUPAC Name (1R,4Z,9S)-4,11,11-trimethyl-8-methylidenebicyclo[7.2.0]undec-4-ene
SMILES C\C\1=C\CCC(=C)[C@H]2CC(C)(C)[C@@H]2CC1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C15H24/c1-11-6-5-7-12(2)13-10-15(3,4)14(13)9-8-11/h6,13-14H,2,5,7-10H2,1,3-4H3/b11-6-/t13-,14-/m1/s1
Formula Weight 204.1878
Chemical Name or Material (-)-Isocaryophyllene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.