missing translation for 'onlineSavingsMsg'
Learn More

Isocarbostyril, 98%

Product Code. 11906501 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11906501

Brand: Thermo Scientific Acros 166460050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 491-30-5 | C9H7NO | 145.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 491-30-5
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
InChI Key VDBNYAPERZTOOF-UHFFFAOYSA-N
Synonym 1-hydroxyisoquinoline, isoquinolin-1 2h-one, isocarbostyril, 1-isoquinolinol, 1 2h-isoquinolinone, isoquinolin-1-ol, isoquinolin-1-one, 1 2h-isoquinolone, 1-hydroxyisoquinolin, isoquinolinol
PubChem CID 10284
ChEBI CHEBI:18350
IUPAC Name 2H-isoquinolin-1-one
SMILES C1=CC=C2C(=C1)C=CNC2=O

Specifications

Melting Point 211.0°C to 214.0°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 21,1
Formula Weight 145.16
Percent Purity 98%
Physical Form Crystalline Powder or Powder
Chemical Name or Material Isocarbostyril
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.