missing translation for 'onlineSavingsMsg'
Learn More

Isocarbofos D7 (isopropyl D7), TRC

Product Code. 30403820
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30403820 10 mg 10mg
30399398 5 mg -
30393983 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30403820 Supplier Toronto Research Chemicals Supplier No. TRCI92352110MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C11H9D7NO4PS
Molecular Weight (g/mol) 296.33
Synonym Isocarbophos D7 (isopropyl D7)
SMILES [2H]C([2H])([2H])C([2H])(OC(=O)c1ccccc1OP(=S)(N)OC)C([2H])([2H])[2H]

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C11H16NO4PS/c1-8(2)15-11(13)9-6-4-5-7-10(9)16-17(12,18)14-3/h4-8H,1-3H3,(H2,12,18)/i1D3,2D3,8D
Chemical Name or Material Isocarbofos D7 (isopropyl D7)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.