missing translation for 'onlineSavingsMsg'
Learn More

Isocapric Acid, TRC

Product Code. 30393513
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30393513 1 g
30401781 100 mg
30388855 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393513 Supplier Toronto Research Chemicals Supplier No. TRCI7892501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5963-14-4
Molecular Formula C10 H20 O2
Molecular Weight (g/mol) 172.26
Synonym 8-Methylnonanoic Acid
IUPAC Name 8-methylnonanoic acid
SMILES CC(C)CCCCCCC(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H20O2/c1-9(2)7-5-3-4-6-8-10(11)12/h9H,3-8H2,1-2H3,(H,11,12)
Formula Weight 172.1463
Chemical Name or Material Isocapric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.