missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ISOC2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27082-1-AP-20UL
This item is not returnable.
View return policy
Description
ISOC2 gene ontology annotations related to this gene include protein destabilization.Specifications
| ISOC2 | |
| Polyclonal | |
| Unconjugated | |
| ISOC2 | |
| ISOC2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 664994, 67441, 79763 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P85094, Q96AB3, Q9DCC7 | |
| ISOC2, Isoc2a, Isoc2b | |
| ISOC2 Fusion Protein Ag23936 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction